Imiquimod 99%

Supplier: Ambeed
Danger

A161049-10G A161049-1G A161049-5G
76824-076EA 106 USD
76824-076 76824-078 76824-080 76824-082
Imiquimod 99%
Imiquimod

Caution: for Research Use Only

Formula: C₁₄H₁₆N₄
MDL Number: MFCD00866946
CAS Number: 99011-02-6
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR