Quercetin dihydrate 95%
Supplier: Ambeed
|
Danger
|
Synonyms:
2-(3,4-Dihydroxyphenyl)-3,5,7-trihydroxy-4H-chromen-4-one dihydrate
A114540-10G
A114540-5G
76822-054EA
10
USD
76822-054
76822-056
Quercetin dihydrate 95%
Quercetin dihydrate
Caution: for Research Use Only
Formula:
C₁₅H₁₄O₉ MW: 338.268 g/mol |
MDL Number:
MFCD00149487 CAS Number: 6151-25-3 UN: 2811 ADR: 6.1 Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...