Dimethyl-2-aminoterephthalate 98%

Supplier: Ambeed
Warning

Synonyms: Dimethyl aminoterephthalate, 2-Aminodimethyl terephthalate

A869436-100G A869436-25G A869436-5G
76820-806EA 80 USD
76820-806 76820-808 76820-810
Dimethyl-2-aminoterephthalate 98%
Dimethyl 2-aminoterephthalate

Caution: for Research Use Only

Formula: C₁₀H₁₁NO₄
MDL Number: MFCD00008427
CAS Number: 5372-81-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR