1-Docosanol 95% (mixture of isomers)
Supplier: Ambeed
|
Warning
|
Synonyms:
Behenyl alcohol
A762512-500G
A762512-1G
A762512-25G
A762512-5G
A762512-100G
A762512-1000G
76820-366EA
10.61
USD
76820-366
76820-368
76820-362
76820-364
76820-360
1-Docosanol 95% (mixture of isomers)
1-Docosanol
Caution: for Research Use Only
Formula:
H₃C(CH₂)₂₀CH₂OH MW: 326.61 g/mol |
MDL Number:
MFCD00002939 CAS Number: 661-19-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...