N2-Isobutyryl-2'-deoxyguanosine 98%

Supplier: Ambeed
Warning

A538127-100MG A538127-10G A538127-1G A538127-1MG A538127-25G A538127-100G A538127-5G A538127-250MG
76818-976EA 6 USD
76818-976 76818-978 76818-980 76818-982 76818-984 77673-440 77673-442 77884-337
N2-Isobutyryl-2'-deoxyguanosine 98%
N2-Isobutyryl-2'-deoxyguanosine

Caution: for Research Use Only

Formula: C₁₄H₁₉N₅O₅
MDL Number: MFCD00010060
CAS Number: 68892-42-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR