Isonicotinaldehyde oxime 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
Pyridine-4-carboxaldoxime
, 4-Pyridinecarbaldehyde oxime
A496951-5G
A496951-25G
76818-720EA
10.3
USD
76818-720
76818-718
Isonicotinaldehyde oxime 98%
Isonicotinaldehyde oxime
Caution: for Research Use Only
Formula:
C₆H₆N₂O MW: 122.13 g/mol |
MDL Number:
MFCD00006446 CAS Number: 696-54-8 UN: 1325 ADR: 4.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...