4-Chloro-3-iodophenol 98%

Supplier: Ambeed
Danger

A435245-1G
76818-186EA 36.05 USD
76818-186
4-Chloro-3-iodophenol 98%
4-Chloro-3-iodophenol

Caution: for Research Use Only

Formula: C₆H₄ClIO
MW: 254.45 g/mol
MDL Number: MFCD00070769
CAS Number: 202982-72-7
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR