3-Nitrophthalic acid 98%

Supplier: Ambeed
Warning

A320154-100G A320154-25G A320154-500G A320154-1000G
76817-046EA 6 USD
76817-046 76817-048 76817-050 77672-644
3-Nitrophthalic acid 98%
3-Nitrophthalic acid

Caution: for Research Use Only

Formula: C₈H₅NO₆
MW: 211.13 g/mol
Boiling Pt: 441
MDL Number: MFCD00007138
CAS Number: 603-11-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR