1-(2-Aminoethyl)pyrrolidine 98%

Supplier: Ambeed
Danger

A291884-10G A291884-1G A291884-5G A291884-25G
76816-592EA 15 USD
76816-592 76816-594 76816-596 77550-770
1-(2-Aminoethyl)pyrrolidine 98%
1-(2-Aminoethyl)pyrrolidine

Caution: for Research Use Only

Formula: C₆H₁₄N₂
MDL Number: MFCD00003182
CAS Number: 7154-73-6
UN: 2734
ADR: 8,3,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR