2-Amino-5-methylbenzonitrile 98%

Supplier: Ambeed
Danger

Synonyms: 2-Cyano-4-methylaniline, 4-Amino-3-cyanotoluene, 6-Amino-m-tolunitrile

A254259-250MG A254259-1G A254259-5G
76815-770EA 10 USD
76815-770 77883-690 77883-691
2-Amino-5-methylbenzonitrile 98%
2-Amino-5-methylbenzonitrile

Caution: for Research Use Only

Formula: C₈H₈N₂
MDL Number: MFCD00267506
CAS Number: 5925-93-9
UN: 3439
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR