2,5-Dibromo-4-pyridinylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (2,5-Dibromopyridin-4-yl)boronic acid

A254086-1G A254086-250MG A254086-5G
76815-758EA 40 USD
76815-758 76815-760 76815-762
2,5-Dibromo-4-pyridinylboronic acid 98%
2,5-Dibromo-4-pyridinylboronic acid

Caution: for Research Use Only

Formula: C₅H₄BBr₂NO₂
MDL Number: MFCD11109338
CAS Number: 1031843-77-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR