4-Amino-2-bromobenzonitrile 98%

Supplier: Ambeed
Warning

A189259-100G A189259-10G A189259-1G A189259-25G A189259-5G
76813-848EA 142 USD
76813-848 76813-850 76813-852 76813-854 76813-856
4-Amino-2-bromobenzonitrile 98%
4-Amino-2-bromobenzonitrile

Caution: for Research Use Only

Formula: C₇H₅BrN₂
MDL Number: MFCD03695814
CAS Number: 53312-82-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR