2-Aminothiazole-5-carbaldehyde 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Aminothiazole-5-carboxaldehyde, 2-Amino-1,3-thiazole-5-carboxaldehyde
A160589-10G
A160589-1G
A160589-250MG
A160589-25G
A160589-5G
76812-706EA
93
USD
76812-706
76812-708
76812-710
76812-712
76812-714
2-Aminothiazole-5-carbaldehyde 97%
2-Aminothiazole-5-carbaldehyde
Caution: for Research Use Only
Formula:
C₄H₄N₂OS MW: 128.15 g/mol |
MDL Number:
MFCD02179570 CAS Number: 1003-61-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...