(S)-Benzyl-2-amino-6-(((benzyloxy)carbonyl)amino)hexanoate hydrochloride 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Nepsilon-Benzyloxycarbonyl-L-lysine benzyl ester hydrochloride
, H-Lys(Z)-OBzl·HCl
, H-Lys(Z)-OBzl.HCl
A152058-10G
A152058-1G
A152058-5G
76812-248EA
5.15
USD
76812-248
76812-246
76812-250
(S)-Benzyl-2-amino-6-(((benzyloxy)carbonyl)amino)hexanoate hydrochloride 98%
(S)-Benzyl-2-amino-6-(((benzyloxy)carbonyl)amino)hexanoate hydrochloride
Caution: for Research Use Only
Formula:
C₂₁H₂₆N₂O₄·HCl MW: 406.9 g/mol |
MDL Number:
MFCD00038981 CAS Number: 6366-70-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...