2-Mercapto-1-methylimidazole 99%
Supplier: Ambeed
|
Warning
|
Synonyms:
Thiamazole
, Methimazole
, 2-Mercapto-N-methyl-1H-imidazole
, 2-Mercapto-N-methylimidazole
, 2-Mercapto-1-methyl-1H-imidazole
, 1-Methyl-1H-imidazole-2(3H)-thione
A119279-100G
A119279-1MG
A119279-5G
A119279-25G
76810-370EA
18.54
USD
76810-370
76810-368
76810-366
76810-372
2-Mercapto-1-methylimidazole 99%
2-Mercapto-1-methylimidazole
Caution: for Research Use Only
Formula:
C₄H₆N₂S MW: 114.17 g/mol |
MDL Number:
MFCD00179321 CAS Number: 60-56-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...