2-Chloro-7-nitrobenzo[d]thiazole 97%

Supplier: Ambeed
Warning

A104300-100MG A104300-1G A104300-250MG
76809-476EA 8 USD
76809-476 76809-478 76809-480
2-Chloro-7-nitrobenzo[d]thiazole 97%
2-Chloro-7-nitrobenzo[d]thiazole

Caution: for Research Use Only

Formula: C₇H₃ClN₂O₂S
MDL Number: MFCD09749259
CAS Number: 2942-22-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR