4'-Hydroxypropiophenone 98%

Supplier: Ambeed
Warning

Synonyms: 4’-Hydroxypropiophenone, Paroxypropione, p-Hydroxypropiophenone

A103825-100G A103825-1G A103825-25G A103825-500G A103825-1000G A103825-100MG
76809-440EA 28 USD
76809-440 76809-442 76809-444 76809-446 77610-800 77882-889
4'-Hydroxypropiophenone 98%
4'-Hydroxypropiophenone

Caution: for Research Use Only

Formula: C₉H₁₀O₂
MDL Number: MFCD00002361
CAS Number: 70-70-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR