Synthware Wide Mouth Media Storage Bottles
Supplier: Kemtech America
B140100 B140250 B140500 B141000
76808-810EA 10.97 USD
76808-810 76808-808 76808-798 76808-802
Synthware Wide Mouth Media Storage Bottles
Bottles Glass Bottles
GL 45 wide mouth media bottle constructed of borosilicate glass 3.3 with colored caps.
  • Heavy wall construction
  • Color-coded by size
  • Writing patch and graduations
The wide mouth accommodates various funnels, scoops, and spatulas. Heavy wall borosilicate glass ensures durability and safe storage of all media. Bottles are autoclavable once lid and pour ring are removed.
Certifications: Conforms to USP Type I and ASTM standards. Meet all Class A and Type 1 requirements.
Accessories information: Color-coded lids are supplied with bottles and are also available separately. All lids are interchangeable with all bottle sizes.
Delivery information: Supplied with screw cap and pouring ring.
Packaging: Bottles are packed in bubble wrap or packing paper to ensure safe arrival.