Mouse ADAMTS8 ELISA Kit
Supplier: Antibodies.com LLC
A303457-96
76735-626EA 745 USD
76735-626
Mouse ADAMTS8 ELISA Kit
Assays ELISAs
Mouse ADAMTS8 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse ADAMTS8 in serum, plasma, tissue homogenates, and other biological fluids.
  • Ready-to-use ELISA kit
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%
Mouse ADAMTS8 ELISA kit (A303457) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse ADAMTS8 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for ADAMTS8 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the ADAMTS8 present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated anti-ADAMTS8 antibody, which binds the captured ADAMTS8 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of ADAMTS8 captured in each well. The concentration of ADAMTS8 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.
Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.