Ethyl N-Cyanoacetimidate 97%
Supplier: Ambeed
|
Danger
|
Synonyms:
N-Cyanoethanimidic acid Ethyl ester, N-Cyanoacetimidic acid Ethyl ester, Ethyl N-Cyanoethanimidate, Ethyl N-cyanoacetimidate
A962581-10G
A962581-25G
A962581-5G
76808-130EA
7
USD
76808-130
76808-132
76808-134
Ethyl N-Cyanoacetimidate 97%
Ethyl N-Cyanoacetimidate
Caution: for Research Use Only
Formula:
C₅H₈N₂O |
MDL Number:
MFCD03411888 CAS Number: 1558-82-3 UN: 3272 ADR: 3,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...