Ethyl acetoacetate 96%

Supplier: Ambeed
Warning

Synonyms: Acetoacetic acid ethyl ester, EAA

A899480-1000G A899480-100G A899480-25G A899480-500G
76807-928EA 15 USD
76807-928 76807-930 76807-932 76807-934
Ethyl acetoacetate 96%
Ethyl acetoacetate

Caution: for Research Use Only

Formula: C₆H₁₀O₃
MDL Number: MFCD00009199
CAS Number: 141-97-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR