DSG (Disuccinimidyl glutarate) 97%

Supplier: Ambeed
Warning

A880434-10MG A880434-1MG A880434-250MG A880434-5G A880434-1G A880434-25G A880434-100MG
76807-846EA 6 USD
76807-846 76807-848 76807-850 76807-852 77552-654 77603-202 77627-702
DSG (Disuccinimidyl glutarate) 97%
DSG (Disuccinimidyl glutarate)

Caution: for Research Use Only

Formula: C₁₃H₁₄N₂O₈
MDL Number: MFCD00153597
CAS Number: 79642-50-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR