Dibutyl Itaconate 98% stabilized
Supplier: Ambeed
|
Warning
|
Stabilization:
Stabilised with hydroquinone (HQ)
A694722-1000G
A694722-100G
A694722-25G
A694722-500G
76806-678EA
108
USD
76806-678
76806-680
76806-682
76806-684
Dibutyl Itaconate 98% stabilized
Dibutyl Itaconate
Caution: for Research Use Only
Formula:
C₁₃H₂₂O₄ |
MDL Number:
MFCD00027211 CAS Number: 2155-60-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...