Lactobionic acid 97%

Supplier: Ambeed
Warning

A651883-100G A651883-1G A651883-25G A651883-500G A651883-5G
76806-368EA 54 USD
76806-368 76806-370 76806-372 76806-374 76806-376
Lactobionic acid 97%
Lactobionic acid

Caution: for Research Use Only

Formula: C₁₂H₂₂O₁₂
MDL Number: MFCD00078147
CAS Number: 96-82-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR