(Ethyl)triphenylphosphonium iodide 97%

Supplier: Ambeed
Danger

A613097-25G A613097-5G
76806-066EA 20 USD
76806-066 76806-068
(Ethyl)triphenylphosphonium iodide 97%
(Ethyl)triphenylphosphonium iodide

Caution: for Research Use Only

Formula: C₂₀H₂₀IP
MDL Number: MFCD00040352
CAS Number: 4736-60-1
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR