di-Lithium tetraborate 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
Dilithium tetraborate
, Lithium tetraborate
A610844-25G
76806-062EA
6.37
USD
76806-062
di-Lithium tetraborate 98%
di-Lithium tetraborate
Caution: for Research Use Only
Formula:
Li₂B₄O₇ MW: 169.12 g/mol |
MDL Number:
MFCD00011083 CAS Number: 12007-60-2 UN: 1759 ADR: 8,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...