(+)-Bis(pinanediolato)diboron 98%

Supplier: Ambeed
Warning

Synonyms: (+)-Bis(pinanediolato)diborane, (3aS,3'aS,4S,4'S,6S,6'S,7aR,7'aR)-(+)-Dodecahydro-3a,3'a,5,5,5',5'-hexamethyl-2,2'-bi-4,6-methano-1,3,2-benzodioxaborole, (3aS,3'aS,4S,4'S,6S,6'S,7aR,7'aR)-Dodecahydro-3a,3'a,5,5,5',5'-hexamethyl-2,2'-bi-4,6-methano-1,3,2-benzodioxaborole

A599126-1G A599126-250MG A599126-5G A599126-10G
76805-954EA 32 USD
76805-954 76805-956 77669-588 77880-273
(+)-Bis(pinanediolato)diboron 98%
(+)-Bis(pinanediolato)diboron

Caution: for Research Use Only

Formula: C₂₀H₃₂B₂O₄
MW: 358.088 g/mol
MDL Number: MFCD27664803
CAS Number: 230299-05-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR