Iodoacetamide 98%

Supplier: Ambeed
Danger

A576369-25G A576369-5G A576369-100G A576369-10G
76805-750EA 42 USD
76805-750 76805-752 77602-422 77602-424
Iodoacetamide 98%
Iodoacetamide

Caution: for Research Use Only

Formula: ICH₂CONH₂
MDL Number: MFCD00008028
CAS Number: 144-48-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR