Diethyl (difluoromethyl)phosphonate 98%

Supplier: Ambeed
Danger

Synonyms: Difluoromethylphosphonicaciddiethylester

A542539-1G A542539-25G A542539-5G
76805-386EA 16.98 USD
76805-386 76805-388 76805-390
Diethyl (difluoromethyl)phosphonate 98%
Diethyl (difluoromethyl)phosphonate

Caution: for Research Use Only

Formula: C₅H₁₁F₂O₃P
MW: 188.1096 g/mol
MDL Number: MFCD00192023
CAS Number: 1478-53-1
UN: 1760
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR