Ethyl (methylthio)acetate 95%
Supplier: Ambeed
|
Danger
|
Synonyms:
(Methylthio)acetic ethyl ester, Ethyl 2-(methylthio)acetate
A479732-25G
A479732-5G
76804-816EA
10.61
USD
76804-816
76804-818
76804-820
Ethyl (methylthio)acetate 95%
Ethyl (methylthio)acetate
Caution: for Research Use Only
Formula:
C₅H₁₀O₂S |
MDL Number:
MFCD00009182 CAS Number: 4455-13-4 UN: 3272 ADR: 3,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...