(4aR,5R,6aS,6bR,8aR,10S,12aR,12bR,14bS)-5,10-Dihydroxy-2,2,6a,6b,9,9,12a-heptamethyl-1,2,3,4,4a,5,6,6a,6b,7,8,8a,9,10,11,12,12a,12b,13,14b-icosahydropicene-4a-carboxylic acid 97%

Supplier: Ambeed
Warning

A446246-10MG A446246-1MG A446246-5MG
76804-446EA 33 USD
76804-446 76804-448 76804-450
(4aR,5R,6aS,6bR,8aR,10S,12aR,12bR,14bS)-5,10-Dihydroxy-2,2,6a,6b,9,9,12a-heptamethyl-1,2,3,4,4a,5,6,6a,6b,7,8,8a,9,10,11,12,12a,12b,13,14b-icosahydropicene-4a-carboxylic acid 97%
(4aR,5R,6aS,6bR,8aR,10S,12aR,12bR,14bS)-5,10-Dihydroxy-2,2,6a,6b,9,9,12a-heptamethyl-1,2,3,4,4a,5,6,6a,6b,7,8,8a,9,10,11,12,12a,12b,13,14b-icosahydropicene-4a-carboxylic acid

Caution: for Research Use Only

Formula: C₃₀H₄₈O₄
MDL Number: MFCD00017295
CAS Number: 510-30-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR