Hexaphenylbenzene 95%

Supplier: Ambeed
Warning

A413175-5G A413175-1G A413175-250MG
76804-074EA 175 USD
76804-074 77879-908 77879-909
Hexaphenylbenzene 95%
Hexaphenylbenzene

Caution: for Research Use Only

Formula: C₄₂H₃₀
MDL Number: MFCD00003057
CAS Number: 992-04-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR