(1,5-Cyclooctadiene)iridium(I)-2,4-pentanedionate 98%

Supplier: Ambeed
Warning

Synonyms: Ir(acac)(COD), (Acetylacetonato)(1,5-cyclooctadiene)iridium(I), Acetylacetonato(1,5-cyclooctadiene)iridium(I), 1,5-Cyclooctadiene(acetylacetonato)iridium(I), Ir(acac)(cod)

A390182-100MG A390182-10G A390182-1G A390182-250MG A390182-5G
76803-840EA 43 USD
76803-840 76803-842 76803-844 76803-846 76803-848
(1,5-Cyclooctadiene)iridium(I)-2,4-pentanedionate 98%
(1,5-Cyclooctadiene)iridium(I)-2,4-pentanedionate

Caution: for Research Use Only

Formula: C₁₃H₁₉O₂Ir
MDL Number: MFCD08273779
CAS Number: 12154-84-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR