1,6-Dibromo-2-naphthol 97%

Supplier: Ambeed
Warning

Synonyms: 1,6-Dibromonaphthalen-2-ol1,6-Dibromo-2-hydroxynaphthalene

A386305-10G A386305-100G A386305-25G A386305-5G
76803-770EA 92.3 USD
76803-770 76803-776 76803-772 76803-774
1,6-Dibromo-2-naphthol 97%
1,6-Dibromo-2-naphthol

Caution: for Research Use Only

Formula: C₁₀H₆Br₂O
MW: 301.97 g/mol
MDL Number: MFCD00003870
CAS Number: 16239-18-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR