HA14-1 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl-2-amino-6-bromo-4-(1-cyano-2-ethoxy-2-oxoethyl)-4H-chromene-3-carboxylate, rel-(R)-Ethyl 2-amino-6-bromo-4-((R)-1-cyano-2-ethoxy-2-oxoethyl)-4H-chromene-3-carboxylate(relative)

A350244-10MG A350244-1MG A350244-50MG A350244-100MG A350244-250MG
76803-278EA 38 USD
76803-278 76803-280 76803-282 77668-606 77668-608
HA14-1 98%
HA14-1

Caution: for Research Use Only

Formula: C₁₇H₁₇BrN₂O₅
MW: 409.24 g/mol
MDL Number: MFCD00218213
CAS Number: 65673-63-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR