Ethyl-trans-2-decenoate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl trans-2-decenoate, trans-2-Decenoic Acid Ethyl Ester

A324542-1G A324542-5G A324542-25G A324542-100G
76802-946EA 10 USD
76802-946 76802-948 77601-454 77668-452
Ethyl-trans-2-decenoate 98%
Ethyl-trans-2-decenoate

Caution: for Research Use Only

Formula: C₁₂H₂₂O₂
MW: 198.31 g/mol
MDL Number: MFCD00015299
CAS Number: 7367-88-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR