Etofibrate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-[[2-(4-Chlorophenoxy)-2-methylpropanoyl]oxy]ethyl Nicotinate, Nicotinic acid 2-[[2-(4-Chlorophenoxy)-2-methylpropanoyl]oxy]ethyl ester, 2-((2-(4-Chlorophenoxy)-2-methylpropanoyl)oxy)ethyl nicotinate, 3-Pyridinecarboxylic acid 2-[2-(4-Chlorophenoxy)-2-methyl-1-oxopropoxy]ethyl ester, 2-[2-(4-Chlorophenoxy)-2-methyl-1-oxopropoxy]ethyl 3-Pyridinecarboxylate
A307383-10MG
A307383-1MG
A307383-25MG
A307383-250MG
A307383-100MG
A307383-1G
76802-678EA
10.61
USD
76802-678
76802-680
76802-682
77601-386
77668-356
77668-358
Etofibrate 98%
Etofibrate
Caution: for Research Use Only
Formula:
C₁₈H₁₈ClNO₅ MW: 363.79 g/mol |
MDL Number:
MFCD00865727 CAS Number: 31637-97-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...