4-(Ethoxycarbonyl)phenylboronic acid pinacol ester 95%

Supplier: Ambeed
Warning

A284200-10G A284200-25G A284200-1G
76802-268EA 136.86 USD
76802-268 76802-270 77668-256
4-(Ethoxycarbonyl)phenylboronic acid pinacol ester 95%
4-(Ethoxycarbonyl)phenylboronic acid pinacol ester

Caution: for Research Use Only

Formula: C₁₅H₂₁BO₄
MDL Number: MFCD03453659
CAS Number: 195062-62-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR