1-Adamantanemethylamine 98%

Supplier: Ambeed
Danger

Synonyms: Adamantan-1-ylmethanamine, 1-Aminomethyladamantane

A265820-1G A265820-250MG A265820-25G A265820-5G
76801-890EA 12.74 USD
76801-890 76801-892 76801-894 76801-896 76801-898
1-Adamantanemethylamine 98%
1-Adamantanemethylamine

Caution: for Research Use Only

Formula: C₁₁H₁₉N
MW: 165.28 g/mol
MDL Number: MFCD00074750
CAS Number: 17768-41-1
UN: 2735
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR