Magnesium acetate tetrahydrate 99%

Supplier: Ambeed
Warning

Synonyms: Acetic acid magnesium salt tetrahydrate, Magnesium di(acetate) tetrahydrate

A264999-1000G A264999-100G A264999-500G
76801-844EA 17 USD
76801-844 76801-846 76801-848
Magnesium acetate tetrahydrate 99%
Magnesium acetate tetrahydrate

Caution: for Research Use Only

Formula: (H₃CCOO)₂Mg·4H₂O
MDL Number: MFCD00149214
CAS Number: 16674-78-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR