D-(+)-Maltose monohydrate 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Maltobiose hydrate, Malt Sugar, beta-Maltose monohydrate, Maltose monohydrate, (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal hydrate, 4-O-α-D-Glucopyranosyl-D-glucose hydrate
A250654-1000G
A250654-100G
A250654-25G
A250654-500G
76801-544EA
92
USD
76801-544
76801-546
76801-548
76801-550
D-(+)-Maltose monohydrate 95%
D(+)-Maltose monohydrate
Caution: for Research Use Only
Formula:
C₁₂H₂₂O₁₁·1H₂O MW: 360.32 g/mol |
MDL Number:
MFCD00149343 CAS Number: 6363-53-7 Merck Index: 13,05736 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...