Imidazolidinyl urea 98%

Supplier: Ambeed
Warning

A204362-1000G A204362-100G A204362-10G A204362-500G A204362-25G
76800-472EA 173 USD
76800-472 76800-474 76800-476 76800-478 76800-480 77086-372
Imidazolidinyl urea 98%
Imidazolidinyl urea

Caution: for Research Use Only

Formula: C₁₁H₁₆N₈O₈
MDL Number: MFCD00221482
CAS Number: 39236-46-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR