DMOG 98%

Supplier: Ambeed
Danger

Synonyms: N-[Methoxy(oxo)acetyl]glycine methyll ester, Dimethyloxaloylglycine, Methyl 2-((2-methoxy-2-oxoethyl)amino)-2-oxoacetate, DMOG

A160647-10MG A160647-1G A160647-1MG A160647-50MG A160647-100MG A160647-250MG A160647-5G
76799-004EA 29 USD
76799-004 76799-006 76799-008 76799-010 77600-274 77600-276 77625-552
DMOG 98%
DMOG

Caution: for Research Use Only

Formula: C₆H₉NO₅
MDL Number: MFCD05865098
CAS Number: 89464-63-1
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR