Ipriflavone 98%

Supplier: Ambeed
Warning

A150093-1000G A150093-100G A150093-100MG A150093-1G A150093-250MG A150093-25G A150093-500G A150093-50MG A150093-5G
76798-596EA 1299 USD
76798-596 76798-598 76798-600 76798-602 76798-604 76798-606 76798-608 76798-610 76798-612
Ipriflavone 98%
Ipriflavone

Caution: for Research Use Only

Formula: C₁₈H₁₆O₃
MDL Number: MFCD00221719
CAS Number: 35212-22-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR