Dioctyltin oxide 98%

Supplier: Ambeed
Warning

Synonyms: Di-n-octyltin oxide, Dioctyloxostannane

A148780-100G A148780-25G
76798-538EA 26 USD
76798-538 76798-540
Dioctyltin oxide 98%
Dioctyltin oxide

Caution: for Research Use Only

Formula: C₁₆H₃₄OSn
MDL Number: MFCD00013839
CAS Number: 870-08-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR