Glutaric acid 95%

Supplier: Ambeed
Warning

A124849-1000G A124849-100G A124849-25G A124849-500G
76797-616EA 48 USD
76797-616 76797-618 76797-620 76797-622
Glutaric acid 95%
Glutaric acid

Caution: for Research Use Only

Formula: C₅H₈O₄
MDL Number: MFCD00004410
CAS Number: 110-94-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR