Indigo carmine 85%
Supplier: Ambeed
|
Warning
|
Synonyms:
CI 73015
, 5,5'-Indigodisulfonic acid, Disodium salt
, Carmine Blue
, Food blue No. 2
, Sodium 3,3'-dioxo-[2,2'-biindolinylidene]-5,5'-disulphonate
, Sodium 3,3'-dioxo-[2,2'-biindolinylidene]-5,5'-disulfonate
, acid Blue 74
, 5,5'-Indigodisulphonic acid, Disodium salt
, FD and C Blue No. 2
, FD&C Blue #2
A123673-25G
A123673-100G
A123673-5G
A123673-500G
76797-540EA
123.6
USD
76797-540
76797-538
76797-536
77592-412
Indigo carmine 85%
Indigo carmine
Caution: for Research Use Only
Formula:
C₁₆H₈N₂Na₂O₈S₂ MW: 466.36 g/mol |
MDL Number:
MFCD00005723 CAS Number: 860-22-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...