(±)-Linalool 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
3,7-Dimethylocta-1,6-dien-3-ol
, (±)-3,7-Dimethyl-1,6-octadien-3-ol
A117669-25G
A117669-500G
A117669-100G
A117669-10G
76797-166EA
5.31
USD
76797-166
76797-164
76797-168
76797-170
(±)-Linalool 97%
(±)-Linalool
Caution: for Research Use Only
Formula:
C₁₀H₁₈O MW: 154.25 g/mol |
MDL Number:
MFCD00008906 CAS Number: 78-70-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...