6-Methylquinaldine 98%

Supplier: Ambeed
Warning

A117201-100G A117201-1G A117201-25G A117201-5G
76797-130EA 172 USD
76797-130 76797-132 76797-134 76797-136
6-Methylquinaldine 98%
6-Methylquinaldine

Caution: for Research Use Only

Formula: C₁₁H₁₁N
MDL Number: MFCD00006762
CAS Number: 877-43-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR