Diethyl (2-oxopropyl)phosphonate 97%

Supplier: Ambeed
Danger

Synonyms: Diethyl acetylmethylphosphonate

A108705-100G A108705-10G A108705-1G A108705-5G A108705-25G
76796-660EA 243 USD
76796-660 76796-662 76796-664 76796-666 77549-094
Diethyl (2-oxopropyl)phosphonate 97%
Diethyl (2-oxopropyl)phosphonate

Caution: for Research Use Only

Formula: C₇H₁₅O₄P
MDL Number: MFCD00044728
CAS Number: 1067-71-6
UN: 3082
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR